Assay Details
Target Gene Details
Entrez Gene ID: | 52589 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
AI848120, D030020D09Rik, D15Ertd412e |
Location: |
Chr.15:37366175-37792519 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncald | NM_001170866.1 | 6 | 739 | NP_001164337.1 |
| NM_001170867.1 | 4 | 673 | NP_001164338.1 | |
| NM_001170868.1 | 3 | 621 | NP_001164339.1 | |
| NM_134094.4 | 6 | 929 | NP_598855.2 | |
| AI893360.1 | 6 | 734 | ||
| AK034031.1 | 4 | 652 | BAC28553.1 | |
| AK048440.1 | 6 | 909 | BAC33338.1 | |
| AK167313.1 | 3 | 591 | BAE39416.1 | |
| AK182572.1 | ||||
| BC011162.1 | 3 | 616 | AAH11162.1 | |
| BC026979.1 | 3 | 575 | AAH26979.1 |

Genomic Map