Assay Details
Target Gene Details
Entrez Gene ID: | 18591 |
Gene Name: | platelet derived growth factor, B polypeptide |
Gene Aliases: |
PDGF-B, Sis |
Location: |
Chr.15:79995865-80014808 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfb | NM_011057.3 | 4 | 1118 | NP_035187.2 |
| AF162784.1 | 4 | 308 | AAD43587.1 | |
| BC023427.1 | 3 | 242 | AAH23427.1 | |
| BC053430.1 | 4 | 1118 | AAH53430.1 | |
| BC064056.1 | 4 | 785 | AAH64056.1 |
More Information
Set Membership: |
|

Genomic Map