Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mov10l1 | NM_031260.2 | 22 | 3162 | NP_112550.2 |
AF285587.1 | 21 | 3051 | AAK31966.1 | |
AF340211.1 | 8 | 1130 | AAK77049.1 | |
AK084786.1 | 6 | 732 | BAC39279.1 | |
AY303754.1 | 6 | 800 | AAP60176.1 | |
JF319145.1 | 22 | 3191 |
