Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mov10l1 | NM_031260.2 | 4 | 466 | NP_112550.2 |
AF285587.1 | 3 | 355 | AAK31966.1 | |
AK131735.1 | 4 | 552 | ||
BG086707.2 | 4 | 466 | ||
JF319145.1 | 4 | 495 |
