Assay Details
Target Gene Details
Entrez Gene ID: | 78757 |
Gene Name: | RPTOR independent companion of MTOR, complex 2 |
Gene Aliases: |
4921505C17Rik, 6030405M08Rik, AVO3, AW492497, D530039E11Rik |
Location: |
Chr.15:6708349-6800401 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rictor | NM_030168.3 | 3 | 167 | NP_084444.3 |
AK019537.1 | 3 | 165 | BAB31784.1 | |
AK029196.1 | 3 | 169 | BAC26344.1 | |
AK036149.1 | 3 | 168 | BAC29321.2 | |
AK036492.1 | 3 | 168 | BAC29451.1 | |
AK054119.1 | 3 | 157 | BAC35663.1 | |
AK082586.1 | 3 | 168 | ||
AK087858.1 | 3 | 169 | BAC40027.1 | |
AY497009.1 | 3 | 145 | AAR89074.1 | |
AY540053.1 | 3 | 165 | AAS46920.1 | |
BC120903.1 | ||||
BC120904.1 | 3 | 165 |
