Assay Details
Target Gene Details
Entrez Gene ID: | 116838 |
Gene Name: | regulating synaptic membrane exocytosis 2 |
Gene Aliases: |
2810036I15Rik, AW048769, RIM2, Rab3ip2, Rim2(+40A), Rim2(+44A), Rim2(+4A), Serg2, Syt3-rs, mKIAA0751 |
Location: |
Chr.15:39198064-39684375 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rims2 | NM_001256382.1 | 11 | 2474 | NP_001243311.1 |
| NM_001256383.1 | 11 | 2474 | NP_001243312.1 | |
| NM_053271.2 | 13 | 2594 | NP_444501.1 | |
| AB021131.1 | 13 | 2548 | BAB18975.1 | |
| AK129206.1 | 8 | 1560 | BAC98016.1 | |
| BC118979.2 | 11 | 2422 | ||
| HM015529.1 | 11 | 2397 |
More Information
Set Membership: |
|

Genomic Map