Assay Details
Target Gene Details
Entrez Gene ID: | 70356 |
Gene Name: | suppression of tumorigenicity 13 |
Gene Aliases: |
1110007I03Rik, 3110002K08Rik, AW555194, HIP, HOP, HSPABP, HSPABP1, PRO0786, SNC6, p48 |
Location: |
Chr.15:81365044-81399694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St13 | NM_133726.2 | NP_598487.1 | ||
| AK013981.1 | ||||
| AK146528.1 | BAE27237.1 | |||
| AK152270.1 | BAE31086.1 | |||
| AK168557.1 | BAE40430.1 | |||
| AK176982.1 | ||||
| BC003843.1 | AAH03843.1 | |||
| CT010318.1 | CAJ18526.1 |

Genomic Map