Assay Details
Target Gene Details
Entrez Gene ID: | 671535 |
Gene Name: | poly (ADP-ribose) polymerase family, member 10 |
Gene Aliases: |
- |
Location: |
Chr.15:76232995-76243440 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp10 | NM_001163575.1 | NP_001157047.1 | ||
| NM_001163576.1 | NP_001157048.1 | |||
| AK028373.1 | BAC25913.1 | |||
| AK035309.1 | ||||
| AK046755.1 | BAC32856.1 | |||
| AK146336.1 | ||||
| AK153205.1 | BAE31805.1 | |||
| AK166291.1 | BAE38685.1 | |||
| BB843953.1 | ||||
| BC024074.1 | AAH24074.1 | |||
| BY742887.1 |

Genomic Map