Assay Details
Target Gene Details
Entrez Gene ID: | 17189 |
Gene Name: | myoglobin |
Gene Aliases: |
AI325109 |
Location: |
Chr.15:77015487-77050668 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mb | NM_001164047.1 | 1 | 166 | NP_001157519.1 |
NM_001164048.1 | NP_001157520.1 | |||
NM_013593.3 | NP_038621.2 | |||
AA271284.1 | ||||
AK137456.1 | BAE23359.1 | |||
BC025172.1 | AAH25172.1 |
