Assay Details
Target Gene Details
Entrez Gene ID: | 223722 |
Gene Name: | malonyl CoA:ACP acyltransferase (mitochondrial) |
Gene Aliases: |
AI225907, BC025519 |
Location: |
Chr.15:83546797-83555757 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcat | NM_001030014.2 | 3 | 684 | NP_001025185.1 |
| AK135457.1 | 3 | 710 | ||
| AK180925.1 | 2 | 221 | ||
| BC025519.1 | 1 | 180 | AAH25519.1 | |
| BC099494.1 | 3 | 684 | AAH99494.1 |

Genomic Map