Assay Details
Target Gene Details
Entrez Gene ID: | 67154 |
Gene Name: | metadherin |
Gene Aliases: |
2610103J23Rik, 3D3, 3D3/Lyric, AEG-1, AV353288, D8Bwg1112e, Lyric |
Location: |
Chr.15:34082719-34143537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtdh | NM_026002.4 | NP_080278.3 | ||
AF501309.1 | AAP30790.1 | |||
AK029915.1 | BAC26673.1 | |||
AK137182.1 | 1 | 3774 | ||
AK151269.1 | BAE30256.1 | |||
AK151810.1 | BAE30709.1 | |||
AK218970.1 | ||||
AY553638.1 | AAS68097.1 | |||
BC023115.1 | ||||
BC036994.1 | AAH36994.1 | |||
BC138858.1 | ||||
BC138859.1 |
