Assay Details
Target Gene Details
Entrez Gene ID: | 67154 |
Gene Name: | metadherin |
Gene Aliases: |
2610103J23Rik, 3D3, 3D3/Lyric, AEG-1, AV353288, D8Bwg1112e, Lyric |
Location: |
Chr.15:34082719-34143537 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtdh | NM_026002.4 | NP_080278.3 | ||
| AF501309.1 | AAP30790.1 | |||
| AK029915.1 | BAC26673.1 | |||
| AK137182.1 | 2 | 4104 | ||
| AK151269.1 | BAE30256.1 | |||
| AK151810.1 | BAE30709.1 | |||
| AK183641.1 | ||||
| AY553638.1 | AAS68097.1 | |||
| BC023115.1 | ||||
| BC036994.1 | AAH36994.1 | |||
| BC138858.1 | ||||
| BC138859.1 |

Genomic Map