Assay Details
Target Gene Details
Entrez Gene ID: | 68501 |
Gene Name: | non-SMC element 2 homolog (MMS21, S. cerevisiae) |
Gene Aliases: |
1110014D18Rik, AI661537 |
Location: |
Chr.15:59374198-59601689 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsmce2 | NM_001164604.1 | NP_001158076.1 | ||
NM_026746.3 | NP_081022.2 | |||
AK003685.1 | BAB22938.1 | |||
AK051277.1 | BAC34589.1 | |||
BC009125.1 | AAH09125.1 | |||
DV057668.1 |
