Assay Details
Target Gene Details
Entrez Gene ID: | 14083 |
Gene Name: | PTK2 protein tyrosine kinase 2 |
Gene Aliases: |
FAK, FRNK, Fadk, mKIAA4203, p125FAK |
Location: |
Chr.15:73205102-73423275 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptk2 | NM_001130409.1 | NP_001123881.1 | ||
NM_007982.2 | NP_032008.2 | |||
AB011499.1 | BAC53890.1 | |||
AB030035.1 | BAC53924.1 | |||
AK079821.1 | BAC37757.1 | |||
AK220543.1 | BAD90317.1 | |||
JN971016.1 | ||||
M95408.1 | AAA37592.1 |
