Assay Details
Target Gene Details
Entrez Gene ID: | 14083 |
Gene Name: | PTK2 protein tyrosine kinase 2 |
Gene Aliases: |
FAK, FRNK, Fadk, mKIAA4203, p125FAK |
Location: |
Chr.15:73205102-73423275 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptk2 | NM_001130409.1 | 19 | 1939 | NP_001123881.1 |
NM_007982.2 | 18 | 1784 | NP_032008.2 | |
AB011499.1 | 19 | 1595 | BAC53890.1 | |
AB030035.1 | BAC53924.1 | |||
AK079821.1 | 19 | 2036 | BAC37757.1 | |
AK220543.1 | 18 | 1784 | BAD90317.1 | |
JN971016.1 | 18 | 1511 | ||
M95408.1 | 17 | 1581 | AAA37592.1 |
