Assay Details
Target Gene Details
Entrez Gene ID: | 140557 |
Gene Name: | structural maintenance of chromosomes 1B |
Gene Aliases: |
SMC-1B, SMC1beta, Smc1l2 |
Location: |
Chr.15:85064689-85131957 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc1b | NM_080470.1 | 3 | 378 | NP_536718.1 |
| AF303827.1 | 3 | 378 | AAL09333.1 | |
| BC129800.1 | 3 | 369 |

Genomic Map