Assay Details
Target Gene Details
Entrez Gene ID: | 223809 |
Gene Name: | submandibular gland protein C |
Gene Aliases: |
2310010P21Rik, DXImx49e, Muc19, Sfc21 |
Location: |
Chr.15:91838328-91861435 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smgc | NM_198927.3 | 14 | 1586 | NP_945121.1 |
| AK009287.1 | 2 | 130 | ||
| AK132579.1 | 13 | 1489 | BAE21241.1 | |
| AY459348.1 | 14 | 1587 | AAR23240.1 | |
| BC038603.1 | 4 | 342 | AAH38603.1 | |
| BC049243.1 | 6 | 591 | AAH49243.1 | |
| BC055490.1 | AAH55490.1 | |||
| BC069958.1 | 5 | 418 | AAH69958.1 | |
| BC092073.1 | AAH92073.1 | |||
| BC138091.1 | 14 | 1587 | ||
| BC145136.1 | 9 | 867 | ||
| EU089958.1 |

Genomic Map