Assay Details
Target Gene Details
Entrez Gene ID: | 30945 |
Gene Name: | ring finger protein 19A |
Gene Aliases: |
AA032313, Dorfin, Rnf19, UIP117, Ubce7ip2, XYbp |
Location: |
Chr.15:36239933-36283723 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf19a | NM_013923.2 | NP_038951.1 | ||
| AF120207.1 | AAF18303.1 | |||
| AK082665.1 | ||||
| AK147768.1 | BAE28125.1 | |||
| BC040769.1 | AAH40769.1 | |||
| CO432840.1 |
More Information
Set Membership: |
|

Genomic Map