Assay Details
Target Gene Details
Entrez Gene ID: | 67959 |
Gene Name: | poly-U binding splicing factor 60 |
Gene Aliases: |
2410104I19Rik, 2810454F19Rik, SIAHBP1 |
Location: |
Chr.15:76070182-76080950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Puf60 | NM_001164600.1 | NP_001158072.1 | ||
| NM_028364.2 | NP_082640.2 | |||
| NM_133691.5 | NP_598452.2 | |||
| AK012434.1 | ||||
| AK075960.1 | ||||
| AK146534.1 | ||||
| AK149632.1 | BAE28998.1 | |||
| AK151526.1 | BAE30474.1 | |||
| AK168789.1 | BAE40622.1 | |||
| AK182693.1 | ||||
| AK188324.1 | ||||
| AK206679.1 | ||||
| AK206684.1 | ||||
| AK207962.1 | ||||
| AK208529.1 | 1 | 59 | ||
| AK208948.1 | ||||
| AK210046.1 | ||||
| AK216996.1 | ||||
| AK218774.1 | ||||
| BC010601.1 | AAH10601.1 |
More Information
Set Membership: |
|

Genomic Map