Assay Details
Target Gene Details
Entrez Gene ID: | 223754 |
Gene Name: | TBC1 domain family, member 22a |
Gene Aliases: |
BC023106, D15Ertd781e |
Location: |
Chr.15:86214421-86498503 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d22a | NM_145476.2 | NP_663451.2 | ||
| AK039627.1 | BAC30403.1 | |||
| AK146872.1 | BAE27495.1 | |||
| AK154226.1 | BAE32447.1 | |||
| AK154602.1 | BAE32705.1 | |||
| AK155459.1 | BAE33274.1 | |||
| AK155770.1 | BAE33429.1 | |||
| BC066009.1 | AAH66009.1 |
More Information
Set Membership: |
|

Genomic Map