Assay Details
Target Gene Details
Entrez Gene ID: | 18256 |
Gene Name: | otoconin 90 |
Gene Aliases: |
Ocn-95, PLA2L, Pla2ll, oc95 |
Location: |
Chr.15:65876050-65912411 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oc90 | NM_010953.2 | NP_035083.1 | ||
| AF091846.1 | AAC99455.1 | |||
| AF093591.1 | AAD08924.1 | |||
| AK012981.1 | BAB28578.1 | |||
| BC048716.1 | AAH48716.1 |
More Information
Set Membership: |
|

Genomic Map