Assay Details
Target Gene Details
Entrez Gene ID: | 20356 |
Gene Name: | sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A |
Gene Aliases: |
5930434A13, 9130201M22Rik, AI464145, Semaf, semF |
Location: |
Chr.15:32244742-32696341 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema5a | NM_009154.2 | NP_033180.2 | ||
| AK043386.1 | BAC31531.1 | |||
| AK046502.1 | ||||
| AK077021.1 | BAC36572.1 | |||
| AK141612.1 | BAE24765.1 | |||
| AK143026.1 | BAE25254.1 | |||
| AK157706.1 | BAE34160.1 | |||
| BC065137.1 | AAH65137.1 | |||
| X97817.1 | CAA66397.1 |
More Information
Set Membership: |
|

Genomic Map