Assay Details
Target Gene Details
Entrez Gene ID: | 12549 |
Gene Name: | Rho GTPase activating protein 31 |
Gene Aliases: |
5830477L08Rik, AU041750, Cdgap, D330026I07Rik |
Location: |
Chr.16:38598343-38713035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap31 | NM_020260.2 | NP_064656.2 | ||
| AF151363.1 | AAD38043.1 | |||
| AK018057.1 | ||||
| AK138417.1 | BAE23651.1 | |||
| AK149975.1 | BAE29205.1 | |||
| AK150109.1 | BAE29314.1 | |||
| AK155729.1 | BAE33406.1 | |||
| AK170685.1 | ||||
| BC138870.1 |
More Information
Set Membership: |
|

Genomic Map