Assay Details
Target Gene Details
Entrez Gene ID: | 11658 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
AI853494, BEN, CD166, DM-GRASP, MuSC, SC1 |
Location: |
Chr.16:52248996-52453255 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alcam | NM_009655.2 | NP_033785.1 | ||
AB003026.1 | BAD18062.1 | |||
AK030851.1 | BAC27159.1 | |||
BC027280.1 | AAH27280.1 | |||
L25274.1 | AAA37528.1 | |||
U95030.1 | AAC06342.1 |
