Assay Details
Target Gene Details
Entrez Gene ID: | 224273 |
Gene Name: | beta-gamma crystallin domain containing 3 |
Gene Aliases: |
5031404N19, AU022790 |
Location: |
Chr.16:59490775-59601399 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crybg3 | NM_174848.3 | NP_777273.2 | ||
| AK030282.1 | BAC26878.1 | |||
| AK162506.1 | BAE36951.1 | |||
| BC043118.1 | AAH43118.1 |

Genomic Map