Assay Details
Target Gene Details
Entrez Gene ID: | 76510 |
Gene Name: | trafficking protein particle complex 9 |
Gene Aliases: |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882 |
Location: |
Chr.15:72589620-73061243 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc9 | NM_001164641.1 | NP_001158113.1 | ||
| NM_001164642.1 | NP_001158114.1 | |||
| NM_029640.2 | NP_083916.1 | |||
| NM_180662.2 | NP_850993.2 | |||
| AK028504.1 | ||||
| AK029106.1 | ||||
| AK138682.1 | BAE23746.1 | |||
| AK156739.1 | BAE33831.1 | |||
| AK173287.1 | BAD32565.1 | |||
| AY630620.1 | AAV31909.1 | |||
| BC034590.1 | AAH34590.1 | |||
| BC070463.1 | AAH70463.1 |

Genomic Map