Assay Details
Target Gene Details
Entrez Gene ID: | 76510 |
Gene Name: | trafficking protein particle complex 9 |
Gene Aliases: |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882 |
Location: |
Chr.15:72589620-73061243 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc9 | NM_029640.2 | 17 | 2106 | NP_083916.1 |
| NM_180662.2 | 17 | 2851 | NP_850993.2 | |
| AK156739.1 | 18 | 2676 | BAE33831.1 | |
| AK173287.1 | 7 | 850 | BAD32565.1 | |
| AK178042.1 | ||||
| AY630620.1 | 17 | 2104 | AAV31909.1 | |
| BC034590.1 | 17 | 2104 | AAH34590.1 | |
| BC070463.1 | 17 | 2851 | AAH70463.1 |

Genomic Map