Assay Details
Target Gene Details
Entrez Gene ID: | 76510 |
Gene Name: | trafficking protein particle complex 9 |
Gene Aliases: |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882 |
Location: |
Chr.15:72589620-73061243 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trappc9 | NM_029640.2 | 19 | 2333 | NP_083916.1 |
NM_180662.2 | 19 | 3078 | NP_850993.2 | |
AK007766.1 | 2 | 113 | BAB25242.1 | |
AK156739.1 | 20 | 2903 | BAE33831.1 | |
AK173287.1 | 9 | 1077 | BAD32565.1 | |
AK177847.1 | 2 | 149 | ||
AK178042.1 | ||||
AY630620.1 | 19 | 2331 | AAV31909.1 | |
BC034590.1 | 19 | 2331 | AAH34590.1 | |
BC070463.1 | 19 | 3078 | AAH70463.1 |
