Assay Details
Target Gene Details
Entrez Gene ID: | 76510 |
Gene Name: | trafficking protein particle complex 9 |
Gene Aliases: |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882 |
Location: |
Chr.15:72589620-73061243 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trappc9 | NM_001164641.1 | 3 | 906 | NP_001158113.1 |
NM_001164642.1 | 3 | 906 | NP_001158114.1 | |
NM_001164643.1 | 3 | 906 | NP_001158115.1 | |
NM_029640.2 | 2 | 134 | NP_083916.1 | |
NM_180662.2 | 3 | 906 | NP_850993.2 | |
AK028504.1 | 3 | 748 | ||
AK029106.1 | 3 | 748 | ||
AK031788.1 | 3 | 754 | BAC27550.1 | |
AK138682.1 | 3 | 753 | BAE23746.1 | |
AK156739.1 | 3 | 731 | BAE33831.1 | |
AY630620.1 | 2 | 132 | AAV31909.1 | |
BC034590.1 | 2 | 132 | AAH34590.1 | |
BC070463.1 | 3 | 906 | AAH70463.1 |
