Assay Details
Target Gene Details
Entrez Gene ID: | 66793 |
Gene Name: | EF hand calcium binding domain 1 |
Gene Aliases: |
4930449A16Rik, 5430404L10Rik |
Location: |
Chr.16:14906257-14969061 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efcab1 | NM_025769.3 | NP_080045.1 | ||
AK015866.1 | BAB30007.1 | |||
AK017267.1 | BAB30660.1 | |||
AK030381.1 | BAC26934.1 | |||
AK038735.1 | BAC30115.1 | |||
AK042748.1 | BAC31349.1 | |||
AK043519.1 | BAC31565.1 | |||
AK077722.1 | BAC36981.1 | |||
BC085167.1 | AAH85167.1 |
