Assay Details
Target Gene Details
Entrez Gene ID: | 385658 |
Gene Name: | neurexophilin and PC-esterase domain family, member 3 |
Gene Aliases: |
Fam55c, Gm1752, Gm611 |
Location: |
Chr.16:55839953-55896773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nxpe3 | NM_001134457.1 | NP_001127929.1 | ||
| AK157499.1 | ||||
| BC150961.1 | ||||
| BC150969.1 |

Genomic Map