Assay Details
Target Gene Details
Entrez Gene ID: | 67367 |
Gene Name: | PAX3 and PAX7 binding protein 1 |
Gene Aliases: |
1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp |
Location: |
Chr.16:91014037-91044734 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paxbp1 | NM_026110.2 | 6 | 1160 | NP_080386.3 |
| AY033907.1 | 6 | 945 | AAK68725.1 | |
| AY033908.1 | 6 | 945 | AAK68726.1 |
More Information
Set Membership: |
|

Genomic Map