Assay Details
Target Gene Details
Entrez Gene ID: | 320214 |
Gene Name: | MYCBP-associated, testis expressed 1 |
Gene Aliases: |
4932425I24Rik, Aat1, Spata26 |
Location: |
Chr.16:38297754-38342253 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Maats1 | NM_001081025.1 | NP_001074494.1 | ||
| AK077038.1 |

Genomic Map