Assay Details
Target Gene Details
Entrez Gene ID: | 212514 |
Gene Name: | spindle and centriole associated protein 1 |
Gene Aliases: |
Ccdc52, D16Ertd480e |
Location: |
Chr.16:44347395-44388492 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spice1 | NM_144550.4 | 15 | 2306 | NP_653133.3 |
| AK040970.1 | 1 | 252 | ||
| AK048789.1 | 15 | 2250 | BAC33458.1 | |
| AY099108.1 | 15 | 2586 | AAM34496.1 | |
| BC052381.1 | 15 | 2260 | AAH52381.1 | |
| BC056960.1 | 15 | 2260 | AAH56960.1 |

Genomic Map