Assay Details
Target Gene Details
Entrez Gene ID: | 212514 |
Gene Name: | spindle and centriole associated protein 1 |
Gene Aliases: |
Ccdc52, D16Ertd480e |
Location: |
Chr.16:44347395-44388492 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spice1 | NM_144550.4 | 2 | 153 | NP_653133.3 |
| AK048789.1 | 2 | 97 | BAC33458.1 | |
| AK165031.1 | BAE38009.1 | |||
| AY099108.1 | 2 | 433 | AAM34496.1 | |
| BC052381.1 | 2 | 107 | AAH52381.1 | |
| BC056960.1 | 2 | 107 | AAH56960.1 | |
| CX568108.1 | 2 | 153 |

Genomic Map