Assay Details
Target Gene Details
Entrez Gene ID: | 74374 |
Gene Name: | C-type lectin domain family 16, member A |
Gene Aliases: |
4932416N17Rik, curt, mKIAA0350 |
Location: |
Chr.16:10545339-10744878 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec16a | NM_001204229.1 | 10 | 1423 | NP_001191158.1 |
| NM_177562.5 | 11 | 1429 | NP_808230.2 | |
| AK089327.1 | 10 | 1353 | ||
| AK122255.1 | 11 | 1381 | BAC65537.1 | |
| AK155314.1 | 10 | 1365 | BAE33184.1 | |
| AK156927.1 | 10 | 1354 | BAE33899.1 | |
| AK170754.1 | 10 | 1354 | BAE42005.1 | |
| AK171134.1 | 10 | 1354 | ||
| BC138907.1 | 11 | 1332 | ||
| BC138909.1 | 11 | 1332 |

Genomic Map