Assay Details
Target Gene Details
Entrez Gene ID: | 117606 |
Gene Name: | biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein |
Gene Aliases: |
4732455C11, mFLJ00376 |
Location: |
Chr.16:44485045-44558870 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Boc | NM_172506.2 | 11 | 2386 | NP_766094.1 |
AF388037.1 | 10 | 1839 | AAK71999.1 | |
AK028770.1 | 11 | 2407 | BAC26110.1 | |
AK028889.1 | 11 | 2387 | BAC26176.1 | |
AK131182.1 | 6 | 4606 | BAD21432.1 | |
AK203190.1 | ||||
BC056138.1 | 2 | 289 | AAH56138.1 | |
BC062892.1 | 11 | 2366 | AAH62892.1 | |
BC078631.1 | 11 | 2330 | AAH78631.1 |
