Assay Details
Target Gene Details
Entrez Gene ID: | 110197 |
Gene Name: | diacylglycerol kinase, gamma |
Gene Aliases: |
2900055E17Rik, 90kDa, AI854428, Dagk3, E430001K23Rik, mKIAA4131 |
Location: |
Chr.16:22466568-22657299 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkg | NM_138650.2 | 2 | 318 | NP_619591.1 |
| AK083258.1 | 2 | 310 | BAC38833.1 | |
| AK087972.1 | 2 | 276 | ||
| AK089261.1 | 2 | 116 | BAC40819.1 | |
| AK220294.1 | 2 | 260 | BAD90219.1 | |
| BY724037.1 | 2 | 318 |
More Information
Set Membership: |
|

Genomic Map