Assay Details
Target Gene Details
Entrez Gene ID: | 14167 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
Location: |
Chr.16:28157783-28753243 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf12 | AF020738.1 | 4 | 597 | AAB71607.1 |
AK011712.2 | 4 | 696 | ||
AK034335.1 | 4 | 555 | BAC28679.1 | |
BB611389.1 | 5 | 562 | ||
BC030485.1 | 5 | 519 | AAH30485.1 | |
CX239635.1 | 5 | 504 | ||
EU234007.1 | 2 | 169 | ||
U66201.1 | 4 | 518 | AAB18917.1 |
