Assay Details
Target Gene Details
Entrez Gene ID: | 117606 |
Gene Name: | biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein |
Gene Aliases: |
4732455C11, mFLJ00376 |
Location: |
Chr.16:44485045-44558870 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Boc | NM_172506.2 | 18 | 3581 | NP_766094.1 |
AF388037.1 | 17 | 3031 | AAK71999.1 | |
AK028770.1 | 18 | 3599 | BAC26110.1 | |
AK028889.1 | 18 | 3582 | BAC26176.1 | |
AK131182.1 | 13 | 5798 | BAD21432.1 | |
AK204390.1 | 3 | 329 | ||
BC026443.1 | 7 | 982 | AAH26443.1 | |
BC056138.1 | 8 | 1928 | AAH56138.1 | |
BC062892.1 | 18 | 3558 | AAH62892.1 | |
BC078631.1 | 18 | 3525 | AAH78631.1 |
