Assay Details
Target Gene Details
Entrez Gene ID: | 12837 |
Gene Name: | collagen, type VIII, alpha 1 |
Gene Aliases: |
Col8a-1 |
Location: |
Chr.16:57624256-57754737 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col8a1 | NM_007739.2 | 2 | 165 | NP_031765.2 |
| AK018742.1 | 2 | 144 | BAB31383.1 | |
| AK029203.1 | 2 | 141 | ||
| AK032048.1 | 2 | 144 | BAC27670.1 | |
| AK046370.1 | 2 | 144 | BAC32693.1 | |
| AK159542.1 | 2 | 140 | BAE35169.1 | |
| BC011061.1 | 2 | 132 | AAH11061.1 | |
| BY289783.1 | 2 | 166 |
More Information
Set Membership: |
|

Genomic Map