Assay Details
Target Gene Details
Entrez Gene ID: | 12524 |
Gene Name: | CD86 antigen |
Gene Aliases: |
B7, B7-2, B7.2, B70, CLS1, Cd28l2, ETC-1, Ly-58, Ly58, MB7, MB7-2, TS/A-2 |
Location: |
Chr.16:36568956-36666077 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd86 | NM_019388.3 | NP_062261.3 | ||
| AF065897.1 | AAD25880.1 | |||
| AF065898.1 | AAD25881.1 | |||
| AF065899.1 | AAD25882.1 | |||
| AF065900.1 | AAD25883.1 | |||
| AK079513.1 | BAC37666.1 | |||
| AK170054.1 | BAE41535.1 | |||
| AK172548.1 | BAE43062.1 | |||
| BC013807.1 | AAH13807.1 | |||
| L25606.1 | AAA79770.1 | |||
| S70108.1 | AAB30744.2 |
More Information
Set Membership: |
|

Genomic Map