Assay Details
Target Gene Details
Entrez Gene ID: | 12846 |
Gene Name: | catechol-O-methyltransferase |
Gene Aliases: |
Comt1, D16Wsu103e, D330014B15Rik |
Location: |
Chr.16:18406882-18426716 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Comt | NM_001111062.1 | NP_001104532.1 | ||
| NM_001111063.1 | NP_001104533.1 | |||
| NM_007744.3 | NP_031770.2 | |||
| AF076156.1 | AAC33334.1 | |||
| AK148311.1 | BAE28473.1 | |||
| BC010402.1 | AAH10402.1 | |||
| CB946852.1 |
More Information
Set Membership: |
|

Genomic Map