Assay Details
Target Gene Details
Entrez Gene ID: | 224170 |
Gene Name: | DAZ interacting protein 3, zinc finger |
Gene Aliases: |
2310047C04Rik, 2A-HUB, 6430549P11Rik, A230104G20 |
Location: |
Chr.16:48924228-48994153 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dzip3 | NM_001110017.1 | 27 | 3349 | NP_001103487.1 |
NM_027341.2 | 26 | 2731 | NP_081617.1 | |
AK122344.1 | 4 | 1892 | BAC65626.1 | |
BC117953.1 | 26 | 2554 | ||
BC117954.1 | 26 | 2560 | ||
EU233997.1 | 19 | 2551 |
