Assay Details
Target Gene Details
Entrez Gene ID: | 13837 |
Gene Name: | Eph receptor A3 |
Gene Aliases: |
AW492086, Cek4, ETK1, End3, Hek, Hek4, Mek4, Tyro4 |
Location: |
Chr.16:63545218-63864157 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha3 | NM_010140.3 | NP_034270.1 | ||
| AK041935.1 | BAC31104.1 | |||
| AK045219.1 | BAC32266.1 | |||
| AK084921.1 | BAC39310.1 | |||
| AK089119.1 | ||||
| AK132335.1 | BAE21112.1 | |||
| BC138641.1 | ||||
| BC138643.1 | ||||
| BC145349.1 | ||||
| M68513.1 | AAA39521.1 | |||
| M68515.1 | AAA39522.1 |
More Information
Set Membership: |
|

Genomic Map