Assay Details
Target Gene Details
Entrez Gene ID: | 14390 |
Gene Name: | GA repeat binding protein, alpha |
Gene Aliases: |
GABPalpha |
Location: |
Chr.16:84834925-84863783 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabpa | NM_008065.2 | NP_032091.2 | ||
| AK139916.1 | BAE24181.1 | |||
| AK145446.1 | BAE26442.1 | |||
| AK145838.1 | BAE26687.1 | |||
| AK148087.1 | BAE28337.1 | |||
| AK150455.1 | BAE29575.1 | |||
| BC013562.1 | AAH13562.1 | |||
| BC052448.1 | AAH52448.1 | |||
| FN545817.1 | ||||
| M74515.1 | AAA53030.1 |
More Information
Set Membership: |
|

Genomic Map