Assay Details
Target Gene Details
Entrez Gene ID: | 14390 |
Gene Name: | GA repeat binding protein, alpha |
Gene Aliases: |
GABPalpha |
Location: |
Chr.16:84834925-84863783 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabpa | NM_008065.2 | 9 | 1478 | NP_032091.2 |
| AK139916.1 | 9 | 1158 | BAE24181.1 | |
| AK145446.1 | 9 | 1158 | BAE26442.1 | |
| AK145838.1 | 9 | 1223 | BAE26687.1 | |
| AK148087.1 | 9 | 1249 | BAE28337.1 | |
| AK150455.1 | 9 | 1158 | BAE29575.1 | |
| AY282796.1 | 2 | 230 | AAQ19481.1 | |
| AY282797.1 | 2 | 230 | AAQ19482.1 | |
| AY282798.1 | 2 | 230 | AAQ19483.1 | |
| AY282799.1 | 2 | 230 | AAQ19484.1 | |
| BC013562.1 | 7 | 873 | AAH13562.1 | |
| BC052448.1 | 9 | 1478 | AAH52448.1 | |
| FN545817.1 | ||||
| M74515.1 | 9 | 1485 | AAA53030.1 |
More Information
Set Membership: |
|

Genomic Map