Assay Details
Target Gene Details
Entrez Gene ID: | 224171 |
Gene Name: | RIKEN cDNA C330027C09 gene |
Gene Aliases: |
AA408511, AU018569, Cip2a, Kiaa1524 |
Location: |
Chr.16:48994188-49019711 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C330027C09Rik | NM_172616.2 | 14 | 1868 | NP_766204.2 |
AK049356.1 | 14 | 1870 | BAC33705.1 | |
AK160639.1 | 14 | 1855 | ||
AK173190.1 | 13 | 1606 | BAD32468.1 | |
BC052662.1 | 5 | 544 | AAH52662.1 | |
BC137719.1 | 14 | 1877 |
