Assay Details
Target Gene Details
Entrez Gene ID: | 224171 |
Gene Name: | RIKEN cDNA C330027C09 gene |
Gene Aliases: |
AA408511, AU018569, Cip2a, Kiaa1524 |
Location: |
Chr.16:48994188-49019711 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C330027C09Rik | NM_172616.2 | NP_766204.2 | ||
AK049356.1 | BAC33705.1 | |||
AK160639.1 | ||||
AK173190.1 | BAD32468.1 | |||
BC052662.1 | AAH52662.1 | |||
BC137719.1 |
