Assay Details
Target Gene Details
Entrez Gene ID: | 224171 |
Gene Name: | RIKEN cDNA C330027C09 gene |
Gene Aliases: |
AA408511, AU018569, Cip2a, Kiaa1524 |
Location: |
Chr.16:48994188-49019711 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C330027C09Rik | NM_172616.2 | 17 | 2166 | NP_766204.2 |
| AK049356.1 | 17 | 2168 | BAC33705.1 | |
| AK160639.1 | 17 | 2153 | ||
| AK173190.1 | 16 | 1904 | BAD32468.1 | |
| BC052662.1 | 8 | 842 | AAH52662.1 | |
| BC137719.1 | 17 | 2175 |

Genomic Map